DuPont™ ProShield® 6 SFR Coveralls with Standard Hood and Attached Socks
Supplier: DuPont
TM122SBUMD002500 TM122SBULG002500 TM122SBUXL002500 TM122SBU2X002500 TM122SBU3X002500 TM122SBU4X002500 TM122SBU5X002500 TM122SBU6X002500
CA13453-000CS 1743.27 CAD
CA13453-000 CA13453-002 CA13453-004 CA13453-006 CA13453-008 CA13453-010 10016-562 10016-564
DuPont™ ProShield® 6 SFR Coveralls with Standard Hood and Attached Socks
Disposable Garments
ProShield® 6 SFR is a cost-efficient disposable overgarment designed to help protect and preserve primary flame resistant garments.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Front zipper closure
  • Serged seams
ProShield® 6 SFR garments are intended to be worn over primary flame resistant garments to provide a barrier against non-hazardous particles and aerosols. ProShield® 6 SFR garments are designed to not contribute to additional burn injury in the event of an industrial fire or electric arc event. ProShield® 6 SFR garments will not ignite and continue to burn after the flame source is removed. ProShield® 6 SFR garments are an excellent choice as a cost efficient cover to protect reusable FR garments. Applications include Steel Mills, Coal Mines, Maintenance Work, Refineries, Laboratories, and Utilities. coveralls feature an attached hood with elastic around the face opening and elastic openings at wrists for a tighter fit. Integrated socks are composed of the same garment material.