DuPont™ ProShield® 6 SFR Coveralls with Laydown Collar
Supplier: DuPont
TM120SBUMD002500 TM120SBULG002500 TM120SBUXL002500 TM120SBU2X002500 TM120SBU3X002500 TM120SBU4X002500 TM120SBU5X002500 TM120SBU6X002500
CA13452-958CS 1294.72 CAD
CA13452-958 CA13452-960 CA13452-962 CA13452-964 CA13452-966 CA13452-968 10016-558 10016-560
DuPont™ ProShield® 6 SFR Coveralls with Laydown Collar
Disposable Garments
ProShield® 6 SFR is a cost-efficient disposable overgarment designed to help protect and preserve primary flame resistant garments.
  • Laydown collar
  • Open wrists and ankles
  • Front zipper closure
  • Serged seams
ProShield® 6 SFR garments are intended to be worn over primary flame resistant garments to provide a barrier against non-hazardous particles and aerosols. ProShield® 6 SFR garments are designed to not contribute to additional burn injury in the event of an industrial fire or electric arc event. ProShield® 6 SFR garments will not ignite and continue to burn after the flame source is removed. ProShield® 6 SFR garments are an excellent choice as a cost efficient cover to protect reusable FR garments. Applications include Steel Mills, Coal Mines, Maintenance Work, Refineries, Laboratories, and Utilities. Coveralls feature a laydown collar and open wrists and ankles.