DuPont™ Tyvek® 400 D Coveralls, Comfort Fit Design
Supplier: DuPont
TD125SWB2X0025CM TD125SWB3X0025CM TD125SWB4X0025CM TD125SWBMD0025CM TD125SWBLG0025CM TD125SWBXL0025CM
10016-534CS 508 CAD
10016-534 10016-536 10016-538 10016-542 CA10016-540 CA10016-544
DuPont™ Tyvek® 400 D Coveralls, Comfort Fit Design
Disposable Garments
DuPont™ Tyvek® 400 D garments combine the protection, durability and comfort of DuPont™ Tyvek® fabric on the front and the comfort, softness and breathability of DuPont™ ProShield® 10 fabric on the back.
  • Comfort fit design
  • Laydown collar
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
  • White front, blue back
The DuPont™ Tyvek® 400 D garments are suitable for applications where comfort requirements are combined with limited protective requirements for frontal exposures. Garments are available in a variety of styles. Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance. Comfort fit design is based on extensive wearer input to provide our most comfortable garment design that enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing, and features an elastic waist to better position the garment. Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls also feature a laydown collar, a longer zipper that extends to chin for complete coverage of neck area, a storm flap made of the same garment material to cover the zipper, elastic openings at wrists and ankles for a tighter fit, and a thumb loop to keep garment sleeve in place.
Certifications: Thickness meets ASTM D1117.