DuPont™ Tyvek® IsoClean® Frocks with Bound Neck and Set Sleeves
Supplier: DuPont
IC270BWHMD00300C IC270BWHLG00300C IC270BWHXL00300C IC270BWH2X00300C IC270BWHSM0030CS IC270BWHMD0030CS IC270BWHLG0030CS IC270BWH2X003000 IC270BWHSM003000 IC270BWH3X00300C IC270BWH3X0030CS IC270BWH4X0030CS IC270BWHMD0030CS IC270BWHSM00300C
CA80076-800CS 3112.27 CAD
CA80076-800 CA80076-802 CA80076-804 CA80076-806 CA80076-846 CA80076-848 CA80076-850 CA80077-056 CA80077-058 CA80077-060 CA80077-062 CA80077-064 CA80077-066 CA80077-074 CA80077-266 CA89129-678 CA89129-680 80076-848 10016-086
DuPont™ Tyvek® IsoClean® Frocks with Bound Neck and Set Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability, and comfort.
  • Bound seams
  • Bound neck
  • Set sleeve design
  • Covered elastic wrists
  • Front snap closure
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Elastic openings at wrists allow for a tighter fit. Bound neck ensures lower particle shedding. Adjustable snap at neck allows for a better fit. Frocks provide body-length jacket design with front snap closures for easy donning and doffing.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners. Clean Processed and Clean Processed, Sterile garments are individually packaged.