DuPont™ Tyvek® IsoClean® Gowns, Knit Cuffs
Supplier: DuPont
Tyvek®
IC701SWH0000 IC701SWH0000
CA80076-996CS 1468.22 CAD
80076-996 CA80076-996
DuPont™ Tyvek® IsoClean® Gowns, Knit Cuffs
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams
  • Bound neck with ties
  • Knit cuffs
  • Bound ties at center front
  • One size fits most
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Secure, knit cuffs ensure a comfortable fit. Bound neck allows for lower particle shedding with ties for an adjustable fit. Bound ties at the waist originate at the front and tie in the back allowing one size to fit most.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.