Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Azure Biosystems
AC2117
10147-310EA 598 CAD
10147-310
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Azure’s high-quality HRP-conjugated secondary antibodies offer unparalleled sensitivity and performance for immunoblotting and ELISA applications.

These secondary HRP conjugated antibodies are optimized to work with Azure’s Radiance and Radiance Plus protein detection systems.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR