Farrior Ear Speculum, OR Grade, Sklar
Supplier: Sklar
67-6043 67-6044 67-6046
10198-436EA 155.1 CAD
10198-436 10198-438 10198-440
Farrior Ear Speculum, OR Grade, Sklar
Specula
Farrior ear speculum set is a cone shaped attachment for an otoscope that allows examination of the ear canal and eardrum.
  • Constructed of non-glare stainless steel
  • Industry standard
The speculum is inserted into the ear canal and the otoscope handle provides a light source and low-power magnification so the practitioner can view the eardrum. Disease and infections of the middle or inner ear can largely be detected by this examination.