Williams Lacrimal Probe, OR Grade, Sklar
Supplier: Sklar
65-4120 65-4121 65-4123 65-4125 65-4127
10197-400EA 687.86 CAD
10197-400 10197-402 10197-404 10197-406 10197-408
Williams Lacrimal Probe, OR Grade, Sklar
Dissection Needles
Sklar's Williams Lachrimal Probe is a very thin malleable rod with a rounded end used in procedures within the lacrimal system, often pertaining to re-establishing tear flow.
This double-ended instrument has different size tips on each end and the center features a tongue tie that is used as a holding point. This device is available in a set of 4 which contains all sizes (1/2, 3/4, 5/6, and 7/8 probes), or in groups of two.