Kogan Endocervical Speculum, OR Grade, Sklar
Supplier: Sklar
90-3997 90-3995 90-3990 90-3994 90-3996 90-3998
10193-338EA 1327.43 CAD
10193-338 10193-356 10197-954 10197-956 10197-958 10197-960
Kogan Endocervical Speculum, OR Grade, Sklar
Specula
Sklar's kogan endocervical speculum is a gynecological instrument used for viewing the distal endocervical canal.
  • Ratchet locking mechanism
  • With narrow blades of 2 to 4 mm wide
  • Serrations outside of the blade
Unlike a traditional duckbill speculum, this single-spring, locking, finger-ring intrument features opposing, fenestrated blades at the end of long jaws. The jaws and blades extend deeper into the cervix and can therefore retract deeper tissue.
An endocervical speculum is frequently used in colposcopy procedures, or where polyps cannot be easily visualized.
Order Now
SPECIFICATIONS
Environmentally Preferable