Buck Neurohammer, OR Grade, Sklar
Supplier: Sklar
06-3370
10192-460EA 102.66 CAD
10192-460
Buck Neurohammer, OR Grade, Sklar
Mallets and Hammers
Buck Neurohammer is a diagnostic tool that features a head design similar to that of the Dejerine Hammer, however the non-latex bumpers on each end of the head are different sizes.
  • Used for reflex and general neurological testing
  • Reusable, latex-free
The Buck Hammer comes with a pin that screws into the top of the handle which protrudes slightly vertically through the middle point of the bumpers. This hammer also contains a brush that screws into the bottom of the handle.