Billeau Ear Curette, OR Grade, Sklar
Supplier: Sklar
67-2522 67-2521 67-2523
10192-298EA 140.08 CAD
10192-298 10192-368 10193-954
Billeau Ear Curette, OR Grade, Sklar
Curettes
Sklar's Billeau Ear Curette is typically used for cleaning out the ear canal.
  • Malleable wire loop end adjusts to the curvature of the ear canal and removes cerumen
  • Available with various loop sizes to accommodate size of the patient's ear
Curette is straight with a sharp, teardrop, fenestrated wire tip.