ESD Safe, Medium Temperature Hot Glove, 14", Transforming Technologies
Supplier: Transforming Technologies
GL9101 GL9102 GL9103 GL9104
10808-984EA 71.6 CAD
10808-984 10808-986 10808-988 10808-990
ESD Safe, Medium Temperature Hot Glove, 14", Transforming Technologies
Gloves
Great static control and heat protection are ensured in one cleanroom compatible glove.
  • These gloves use a polyester knit fabric suffused with carbon fiber
  • Heat protection is enhanced with a 100% inner tricot knit polyester fabric liner and a full 35.6 cm (14") length
  • A cuff mounted grounding snap is also included
Designed for use with printed circuit boards, hybrid circuits, semiconductors, and other processes, the GL9100 series ESD safe hot glove provides static protection without compromising cleanliness. Gloves resist heat up to 150 °C (302 °F).