Sodium chloride, VWR® USP, JP, Ph. Eur., BP endotoxin tested

Supplier: Avantor Performance Materials

Synonyms: Saline, NaCl, Sea salt, Halite

VWRB241-12KG VWRB241-1KG VWRB241-50KG VWRB241-F350LB VWRB241-P350LB
VWRB241-12KGEA 1767.36 CAD
VWRB241-12KG VWRB241-1KG VWRB241-50KG VWRB241-F350LB VWRB241-P350LB
Sodium chloride, VWR® USP, JP, Ph. Eur., BP endotoxin tested
Sodium chloride

Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Acidity or Alkalinity Passes Test
Aluminum ≤ 0.2 ppm
Appearance of Solution Passes Test
Arsenic ≤ 1.0 ppm
Endotoxin ≤ 2.5EU/g
Assay (dried basis) 99.0% - 105%
Barium Test Passes Test
Ferrocyanides Passes Test
Heavy Metals ≤ 3.0 ppm
Identification A Passes Test
Identification B Passes Test
Iodides Passes Test
Iron ≤ 2.0 ppm
Limit of Bromides ≤ 100 ppm
Limit of Phosphates ≤ 25 ppm
Limit of Potassium ≤ 500 ppm
Loss on Drying ≤ 0.5%
Magnesium and Alkaline - Earth Metals ≤ 100 ppm
Nitrites Passes Test
Sulfate ≤ 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR