Anti-CD184 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: L276F12]

Supplier: Biolegend
146503 146504
CA10756-542EA 234.58 CAD
CA10756-542 CA10756-544
Anti-CD184 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: L276F12]
Antibodies
Alexa Fluor® 647 anti-mouse CD184 (CXCR4) [L276F12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD184
Clonality: Monoclonal
Clone: L276F12
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR