Anti-CD183 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: G025H7]

Supplier: Biolegend
353735 353736
CA10759-722EA 244.16 CAD
CA10759-722 CA10759-724
Anti-CD183 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: G025H7]
Antibodies
PE/Dazzle™ 594 anti-human CD183 (CXCR3) [G025H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD183
Clonality: Monoclonal
Clone: G025H7
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR