Anti-CD183 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: G025H7]

Supplier: Biolegend
353731 353732
CA10759-716EA 325.54 CAD
CA10759-716 CA10759-718
Anti-CD183 Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: G025H7]
Antibodies
Brilliant Violet 711™ anti-human CD183 (CXCR3) [G025H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD183
Clonality: Monoclonal
Clone: G025H7
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR