Anti-CD169 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7-239]

Supplier: Biolegend
346007 346008
CA346007-BLEA 218.62 CAD
CA346007-BL CA346008-BL
Anti-CD169 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7-239]
Antibodies
APC anti-human CD169 (Sialoadhesin, Siglec-1) [7-239]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD169
Clonality: Monoclonal
Clone: 7-239
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR