Anti-CD163 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: GHI/61]

Supplier: Biolegend
333617 333618
CA10758-500EA 225.01 CAD
CA10758-500 CA10758-502
Anti-CD163 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: GHI/61]
Antibodies
FITC anti-human CD163 [GHI/61]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: GHI/61
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR