Water USP, purified, not sterile

Supplier: Cytiva

Synonyms: Hydrogen oxide

WPW-USN-1X6 WPW-USN-20L WPW-USN-2XL WPW-USN-1X6 WPW-USN-20L WPW-USN-2XL
10837-208CS 156.42 CAD
10837-208 10837-210 10837-212 CA10837-208 CA10837-210 CA10837-212
Water USP, purified, not sterile
Water

Cytiva's Purified Water, USP can be used as an excipient in the production of non-parenteral preparations such as cleaning of certain equipment and non-parenteral product-contact components.


  • Non-sterile
  • Purified utilizing a nine step process resulting in very high quality water


Available in sterile and non-sterile formats, it complies with the USP monograph for purified water.


Certifications: Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear Liquid (Visual)
Conductivity (Packaged) 5.0 µs/cm (USP 645)
TOC (Before Packaging) 500 ppb (USP 643)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR