Anti-CD158b (KIR2DL2/L3, NKAT2) Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: DX27]

Supplier: Biolegend
312613 312614
CA10758-090EA 280.86 CAD
CA10758-090 CA10758-092
Anti-CD158b (KIR2DL2/L3, NKAT2) Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: DX27]
Antibodies
PerCP/Cy5.5 anti-human CD158b (KIR2DL2/L3, NKAT2) [DX27]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD158b (KIR2DL2/L3, NKAT2)
Clonality: Monoclonal
Clone: DX27
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR