Anti-CD154 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MR1]

Supplier: Biolegend
106511 106512
CA10756-880EA 197.88 CAD
CA10756-880 CA10756-882
Anti-CD154 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MR1]
Antibodies
PE/Cy7 anti-mouse CD154 [MR1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD154
Clonality: Monoclonal
Clone: MR1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR