Anti-CD138 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: MI15]

Supplier: Biolegend
356519 356520
CA10760-168EA 325.54 CAD
CA10760-168 CA10760-170
Anti-CD138 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: MI15]
Antibodies
Brilliant Violet 605™ anti-human CD138 (Syndecan-1) [MI15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD138
Clonality: Monoclonal
Clone: MI15
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR