Anti-CD138 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: MI15]

Supplier: Biolegend
356515 356516
CA10759-432EA 303.2 CAD
CA10759-432 CA10759-434
Anti-CD138 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: MI15]
Antibodies
Brilliant Violet 421™ anti-human CD138 (Syndecan-1) [MI15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD138
Clonality: Monoclonal
Clone: MI15
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD138
Environmentally Preferable
Antigen synonyms B-B4Syndecan1B Cell|SDC1
Antibody type Primary
Clonality Monoclonal
Conjugation Brilliant Violet® 421
Clone MI15
Reactivity Human
Host Mouse
Gene ID 6382
Isotype IgG1 kappa
Immunogen A mixture of U266 and XG-1 human myeloma cell lines.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR