Anti-CD138 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 281-2]

Supplier: Biolegend
142507 142508
CA142507-BLEA 307.99 CAD
CA142507-BL CA142508-BL
Anti-CD138 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 281-2]
Antibodies
Brilliant Violet 421™ anti-mouse CD138 (Syndecan-1) [281-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For flow cytometric staining using the μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD138
Clonality: Monoclonal
Clone: 281-2
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR