Anti-CD135 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: A2F10]

Supplier: Biolegend
135313 135314
CA10759-298EA 290.44 CAD
CA10759-298 CA10759-300
Anti-CD135 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: A2F10]
Antibodies
Brilliant Violet 421™ anti-mouse CD135 [A2F10]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the test sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For flow cytometric staining using the μg size, the suggested use of the this reagent is  0.5 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD135
Clonality: Monoclonal
Clone: A2F10
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR