Anti-CD127 (IL-7Ra) Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: A019D5]

Supplier: Biolegend
351333 351334
CA10759-010EA 360.65 CAD
CA10759-010 CA10759-012
Anti-CD127 (IL-7Ra) Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: A019D5]
Antibodies
Brilliant Violet 605™ anti-human CD127 (IL-7Rα) [A019D5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD127 (IL-7Ra)
Clonality: Monoclonal
Clone: A019D5
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR