Anti-CD127 (IL-7Ra) Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: A019D5]

Supplier: Biolegend
351317 351318
CA351317-BLEA 263.31 CAD
CA351317-BL CA351318-BL
Anti-CD127 (IL-7Ra) Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: A019D5]
Antibodies
Alexa Fluor® 647 anti-human CD127 (IL-7Rα) [A019D5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD127 (IL-7Ra)
Clonality: Monoclonal
Clone: A019D5
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR