Water USP for irrigation (WIR), sterile

Supplier: Cytiva

Synonyms: Hydrogen oxide

WIR-USP-1X6 Z0304 WIR-USP-5XM WIR-USP-1X6 WIR-USP-5XM Z0304
10837-196CS 187.5 CAD
10837-196 10837-198 10837-200 76312-764 76353-870 CA10837-196 CA10837-198 CA10837-200 CA76353-870 CA76312-764
Water USP for irrigation (WIR), sterile
Water

Water for Irrigation, USP grade is a sterile, hypotonic, non-pyrogenic fluid or solvent. It is prepared by terminal distillation and contains no antimicrobial or bacteriostatic agents or added buffers. The pH is 5.0-7.0 and complies with the USP monograph for water for irrigation.


Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.

An excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications. It is also widely used in medical device manufacturing/processing applications where a high quality, sterile water is required

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR