Water USP for injection (WFI), sterile

Supplier: Cytiva

Synonyms: Hydrogen oxide

WFI-USP-1X6 WFI-USP-20L WFI-USP-2XL WFI-USP-1X6 WFI-USP-2XL
10837-190CS 197.72 CAD
10837-190 10837-192 10837-194 75881-010 76207-690 76207-692 CA76207-692 CA10837-190 CA76207-690 CA10837-192 CA10837-194 CA75881-010
Water USP for injection (WFI), sterile
Water

Water for Injection (WFI) USP grade is a terminally distilled, sterile, non-pyrogenic preparation which contains no bacteriostat, antimicrobial agent or added buffer. It complies with the USP monograph for water for injection packaged in bulk for commercial use..


Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.

Water for Injection is an excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications. It is also widely used in medical device manufacturing/processing applications where a high quality, sterile water is required

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR