Anti-CD122 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: TM-beta1]

Supplier: Biolegend
123211 123212
CA10757-028EA 244.16 CAD
CA10757-028 CA10757-030
Anti-CD122 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: TM-beta1]
Antibodies
PerCP/Cy5.5 anti-mouse CD122 (IL-2Rβ) [TM-β1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD122
Clonality: Monoclonal
Clone: TM-beta1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR