Anti-CD11c Armenian Hamster Monoclonal Antibody (Brilliant Violet® 510) [clone: N418]

Supplier: Biolegend
117337 117338
CA10756-366EA 280.86 CAD
CA10756-366 CA10758-872
Anti-CD11c Armenian Hamster Monoclonal Antibody (Brilliant Violet® 510) [clone: N418]
Antibodies
Brilliant Violet 510™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: Brilliant Violet® 510
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR