Anti-CD11c Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 3.9]

Supplier: Biolegend
301633 301634
CA10757-640EA 290.44 CAD
CA10757-640 CA10757-642
Anti-CD11c Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 3.9]
Antibodies
Brilliant Violet 510™ anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: 3.9
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Rhesus;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR