Anti-CD11b Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: ICRF44]

Supplier: Biolegend
301335 301336
CA10757-544EA 333.52 CAD
CA10757-544 CA10757-546
Anti-CD11b Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: ICRF44]
Antibodies
Brilliant Violet 650™ anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Common Marmoset;Cynomolgus;Rhesus;Swine (Pig;Porcine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR