Anti-CD115 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: AFS98]

Supplier: Biolegend
135511 135512
CA135511-BLEA 205.86 CAD
CA135511-BL CA135512-BL
Anti-CD115 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: AFS98]
Antibodies
Alexa Fluor® 488 anti-mouse CD115 (CSF-1R) [AFS98]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD115
Clonality: Monoclonal
Clone: AFS98
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol CD115
Environmentally Preferable
Antigen synonyms M-CSFR|c-fms|CSF-1R|CSF1R
Antibody type Primary
Clonality Monoclonal
Conjugation Alexa Fluor® 488
Clone AFS98
Reactivity Mouse
Host Rat
Gene ID 12978
Isotype IgG2a kappa
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR