Anti-Bcl-6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7D1]

Supplier: Biolegend
358503 358504
CA10760-372EA 244.16 CAD
CA10760-372 CA10760-438
Anti-Bcl-6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7D1]
Antibodies
PE anti-human/mouse Bcl-6 [7D1]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bcl-6
Clonality: Monoclonal
Clone: 7D1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR