Viva Guard Cartridges, Restek
Supplier: Restek
951050210 951050212 951250212 951350210 951350212 951450210 951450212 951650210 951650212 951950210 951950212
10840-366PK 415.55 CAD
10840-366 10840-368 10840-374 10840-380 10840-382 10840-388 10840-390 10840-396 10840-398 10840-404 10840-406
Viva Guard Cartridges, Restek
Chromatography Columns
Viva guard cartridges protect HPLC columns as well as remove particulate contaminants and highly adsorptive compounds from samples.
  • Available in a number of sizes
  • Removes particulate contaminants
Effective for prolonging analytical column life.