Pinnacle® DB Guard Cartridges, Restek
Supplier: Restek
940950210 940950212 941050210 941050212 941350210 941350212 941450210 941450212 941650210 941650212 941850210 941850212 941950210 941950212
10840-316PK 404.37 CAD
10840-316 10840-318 10840-324 10840-326 10840-328 10840-330 10840-332 10840-334 10840-342 10840-344 10840-350 10840-352 10840-358 10840-360
Pinnacle® DB Guard Cartridges, Restek
Chromatography Columns
Pinnacle® DB guard cartridges protect HPLC columns as well as remove particulate contaminants and highly adsorptive compounds from samples, prolonging analytical column life.
  • Available in a number of sizes
  • Removes particulate contaminants
Effective for prolonging analytical column life.