Raptor™ EXP® Guard Column Cartridges, Restek
Supplier: Restek
9309A0252 9309A0253 930450250 930450252 930450253 930950250 930950252 930950253 931450250 931450252 931450253 9304A0250 9304A0252 9304A0253 9309A0250 9314A0250 9314A0252 9314A0253
10060-086PK 1341.42 CAD
10060-086 10060-088 10840-298 10840-300 10840-302 10840-304 10840-306 10840-308 10840-310 10840-312 10840-314 10840-930 10840-932 10840-934 10840-968 10840-988 10840-990 10840-992
Raptor™ EXP® Guard Column Cartridges, Restek
Chromatography Columns
To help protect your investment and further extend the life of Restek's already-rugged LC columns, Restek offers the patent-pending guard column hardware developed by Optimize Technologies.
  • Available in a number of sizes
  • Maximum cartridge pressure: 600 bar/8700 psi (2.7 µm) or 400 bar/5800 psi (5 µm)
A Restek LC guard cartridge in an EXP® direct connect holder is the ultimate in column protection.