Raptor™ Biphenyl LC Columns, Restek
Supplier: Restek
9309A12 9309A15 9309A1E 9309A52 9309A55 9309A5E 9309A65 9309512 9309515 9309552 9309555 9309562 9309565 9309575 930951E 930953E 930955E 930956E 9309A32 9309A35 9309A3E 9309A62 9309A6E
10060-090EA 2278.67 CAD
10060-090 10060-092 10060-094 10060-096 10060-098 10060-100 10060-102 10839-674 10839-676 10839-678 10839-680 10839-682 10839-684 10839-686 10840-960 10840-962 10840-964 10840-966 10840-970 10840-972 10840-974 10840-976 10840-978
Raptor™ Biphenyl LC Columns, Restek
Chromatography Columns
The innovative Biphenyl is Restek's most popular LC stationary phase because it is particularly adept at separating compounds that are hard to resolve or that elute early on C18 and other phenyl chemistries.
  • Increased retention for dipolar, unsaturated, or conjugated solutes
  • Enhanced selectivity when used with methanolic mobile phase
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses
As a result, the rugged Raptor™ Biphenyl column is extremely useful for fast separations in bioanalytical testing applications like drug and metaboliteMore Product Information