Allure® Organic Acids HPLC Columns, Restek
Supplier: Restek
9165585 9165565 9165575 9165565-700 916556E 9165585-700
10803-444EA 2171.9 CAD
10803-444 10839-452 10839-456 10840-642 10840-644 10840-648 10840-650
Allure® Organic Acids HPLC Columns, Restek
Chromatography Columns
Allure Organic Acids columns provide enhanced retention and selectivity for polar organic acids, allowing the separation to be performed on a single 30 cm column.
  • Enhanced retention and selectivity
  • Stable and reproducible
An Allure Organic Acids column effectively resolves key organic acids such as tartaric and quinic acids using the chromatographic conditions specified in AOAC method 986.13. Retention is stable and reproducible, even with the 100% aqueous mobile phase specified in the AOAC method.