Ultra Bulk Packing Materials (5 µm), Restek
Supplier: RESTEK CORP
91005
91015
91025
91035
91065
91075
91745
10839-030EA
1618.72
CAD
10839-030
10839-032
10839-034
10839-036
10839-040
10839-044
10839-046
Ultra Bulk Packing Materials (5 µm), Restek
Chromatography Sorbents
HPLC Sorbents
Restek's extensive QC program ensures the high quality and reproducibility of these silica packing materials.
- Prepare your own columns in conventional or custom dimensions
- Consistent, high-quality materials
Be confident that you are getting consistent premium product. Use these materials for easy scale-up preparative chromatography or for packing your own columns.