Viva C4 Columns, Restek

Supplier: Restek
951251E 9512522-700 951252E 951253E 951255E 9512565-700 951256E 951257E 9512511 9512512 9512515 9512522 9512525 9512532 9512535 9512551 9512552 9512555 9512561 9512565 9512571 9512572
10838-724EA 1029.11 CAD
10838-724 10838-726 10838-728 10838-730 10838-732 10838-734 10838-736 10838-738 10838-740 10838-742 10838-744 10838-746 10838-748 10838-750 10838-752 10838-754 10838-756 10838-758 10838-760 10838-762 10838-764 10838-766 10838-768 10838-770 10840-042 10840-044 10840-046 10840-054 10840-056 10840-058 10840-060 10840-062 10840-064 10840-066 10840-068 10840-070 10840-078 10840-080 10840-088 10840-090
Viva C4 Columns, Restek
Chromatography Columns
Base-deactivated, wide-pore packing exhibits excellent peak shape for a wide range of compounds.

  • Excellent for separating peptides or proteins
  • Rugged, spherical particles with 300 Å pore size
  • High proportion of pore/surface area available to large molecules

Less retention in reversed-phase assays than Viva C18 or Viva C8.

Particle size: 5 µm, spherical
Pore size: 300 Å
Carbon load: 3.5%
End-cap: yes
pH range: 2.5 to 8
Temperature limit: 80 °C
USP phase code: L26
Phase category: C4, butylsilane
Ligand type: monomeric C4
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR