Ultra C1 HPLC Columns (USP L13), Restek
Supplier: Restek
9101565 9101575 910131E 910133E 910135E 910136E 910151E 910152E 910153E 9101555-700 910155E 910156E 910157E 9101312 9101315 9101332 9101335 9101352 9101355 9101362 9101365 9101512 9101515 9101522 9101525 9101532 9101535 9101552 9101555 9101562 9101572
10059-604EA 1153.83 CAD
10059-604 10059-606 10837-882 10837-884 10837-886 10837-888 10837-890 10837-894 10837-896 10837-898 10837-900 10837-902 10837-904 10837-906 10837-908 10837-910 10837-912 10837-914 10837-916 10837-918 10837-920 10837-922 10837-924 10837-926 10837-928 10837-930 10837-934 10837-936 10837-938 10837-940 10837-942 10837-944 10837-946 10837-948 10837-950 10837-952 10837-954 10837-956 10837-958 10837-960 10839-126 10839-128 10839-132 10839-134 10839-138 10839-140 10839-144 10839-146 10839-150 10839-152 10839-156 10839-158 10839-162 10839-164 10839-168 10839-170 10839-174 10839-178
Ultra C1 HPLC Columns (USP L13), Restek
Chromatography Columns
The Ultra line represents Restek’s broadest selection of stationary phases on a single silica support.
  • High-purity silica
  • High-density bonding
  • Designed for selectivity
Made of high-purity, type-B silica that minimizes activity and creates high-density bonding, these columns are designed for selective and reliable HPLC applications.
This exceptionally stable C1 phase features our least-retentive reversed-phase hydrocarbon packing.