Ultra Silica Columns, Restek

Supplier: Restek
910055E 9100562-700 9100565-700 910056E 9100575-700 910057E 9100332 9100335 9100355 9100362 9100365 9100515 9100522 9100525 9100532 9100535 9100552 9100555 9100572 9100312-700 9100315-700 910031E 910033E 9100352-700 910035E 9100365-700 910036E 910051E 910052E 9100535-700 910053E
10837-860EA 908.55 CAD
10837-860 10837-862 10837-864 10837-866 10837-868 10837-870 10837-872 10837-874 10837-876 10837-878 10837-880 10839-062 10839-064 10839-068 10839-072 10839-074 10839-078 10839-086 10839-088 10839-092 10839-094 10839-098 10839-102 10839-118 10840-412 10840-414 10840-416 10840-418 10840-420 10840-424 10840-426 10840-428 10840-430 10840-432 10840-434 10840-436 10840-440 10840-442 10840-444 10840-446 10840-448 10840-450 10840-452 10840-456 10840-458 10840-460 10840-464 10840-466 10840-468
Ultra Silica Columns, Restek
Chromatography Columns
Base-deactivated spherical silica is useful for normal-phase or HILIC separations.

  • Particle Size: 3 or 5 µm, spherical
  • Pore size: 100 Å
  • End-cap: no
  • pH range and temperature limit: 2.5 to 8 and 80 °C
  • USP phase code: L3
  • Phase category: bare silica
  • Ligand type: none

Excellent for separating peptides or proteins, these silica columns have rugged, spherical particles with a pore size of 100 Å. High proportion of pore/surface area available to large molecules.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR