Pinnacle® DB Cyano HPLC Columns (USP L10), Restek
Supplier: Restek
9416552 9416562 9416565 9416575 941656E 941657E 9416512 9416572 941651E 941652E 941653E 941655E 9416575-700 9416515 9416522 9416525 9416532 9416535 9416555
10059-902EA 956.77 CAD
10059-902 10059-904 10059-906 10059-908 10060-132 10060-134 10837-704 10837-706 10838-594 10838-596 10838-598 10838-600 10838-602 10838-604 10838-606 10838-608 10838-610 10838-612 10838-614 10838-616 10838-618 10838-620 10838-622 10838-624 10838-626 10838-628 10838-630 10838-632 10838-634 10838-636 10839-830 10839-840 10839-842 10839-846 10839-848 10839-852
Pinnacle® DB Cyano HPLC Columns (USP L10), Restek
Chromatography Columns
The Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired.
  • Recommended for assays
  • For protonated base analysis
Cyano columns can be used in normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes.
Pinnacle DB columns are 100% manufactured by Restek in our Bellefonte, Pennsylvania facility. Because performance begins with the support, our Pinnacle DB UHPLC columns start with base-deactivated sphericalMore Product Information