Sims-Kelly Retractor, OR Grade, Sklar
Supplier: Sklar
90-3422 90-3430
10199-226EA 1319.78 CAD
10199-226 10199-228
Sims-Kelly Retractor, OR Grade, Sklar
Retractors
The Sklar Sims-Kelly Retractor is used for retracting the vaginal wall during OB/GYN procedures
It is a single-end, right angle, manual retractor available in multiple sizes. This retractor combines multiple characteristics of the standard Kelly and standard Sims retractors. The shaft of this retractor curves backwards slightly before curving forward again. This creates a gentle right angle and a blade that is marginally larger than a standard Kelly retractor, but smaller than a standard Sims retractor. The blade itself has a curvature similar to that of the Sims retractor. The walls are U-shaped, unlike a Kelly retractor. There is no downward turning lip at the tip of the blade as there is with the Kelly retractor, and the tip is squared and blunt, unlike the standard Sims retractor which is rounded.