Sims Vaginal Retractor, Single-Ended, OR Grade, Sklar
Supplier: Sklar
90-3225 90-3230 90-3235 90-3327 90-3330 90-3335
10194-472EA 451.11 CAD
10194-472 10194-474 10194-476 10199-220 10199-222 10199-224
Sims Vaginal Retractor, Single-Ended, OR Grade, Sklar
Retractors
Sklar's Sims Vaginal Retractor is a handheld retractor used primarily for vaginal retraction and exposure
This retractor is a single-ended instrument with the blades always curved at right angles. The handle is not distinctly separate from the working end of the instrument and is essentially a continuous piece of stainless steel. There is a slight curve at the base of the handle for finger placement. The blade is inserted into the vagina to retract the anterior wall so that the posterior wall can be viewed. The blade is trough shaped, which serves to catch and drain blood or fluid that may exit the surgical site.