TitraLab® AT1000 Potentiometric Titrator, 2 Burettes, 2 Pumps
Supplier: Hach
AP0005.AT1222 AP0011.AT1222 AP0013.AT1222 AT1222.97
10662-172EA 6268.51 CAD
10662-172 10662-200 10662-204 10662-258 CA10662-172 CA10662-200 CA10662-204 CA10662-258
TitraLab® AT1000 Potentiometric Titrator, 2 Burettes, 2 Pumps
Titrators
Automatic titration without all the complications.
  • Reliable titration results
  • Eliminates operator interpretation and manual processes
  • Simple setup and titration
  • Application-specific functions
Simplify complex titration with the TitraLab® AT1000 from Hach.
Eliminates operator interpretation and manual processes with automatic titration to quickly deliver accurate and repeatable results. Pre-programmed titration methods detect end points and eliminate manual calculations to make results easier to achieve without advanced programming. Application-More Product Information