TitraLab® AT1000 Potentiometric Titrator, 1 Burette, 2 Pumps
Supplier: Hach
AP0007.AT1122 AP0012.AT1122 AT1122.97
10662-156EA 3312.98 CAD
10662-156 10662-202 10662-256 CA10662-156 CA10662-202 CA10662-256
TitraLab® AT1000 Potentiometric Titrator, 1 Burette, 2 Pumps
Titrators
Automatic titration without all the complications.
  • Reliable titration results
  • Eliminates operator interpretation and manual processes
  • Simple setup and titration
  • Application-specific functions
Simplify complex titration with the TitraLab® AT1000 from Hach.
Eliminates operator interpretation and manual processes with automatic titration to quickly deliver accurate and repeatable results. Pre-programmed titration methods detect end points and eliminate manual calculations to make results easier to achieve without advanced programming. Application-specific functions eliminate complex titration setup and analysis. Hach’s unique application kits make it quick for anyone to set up and operate a test.
Recommended Application Packages include Amperometric: Free and total chlorine, chlorine dioxide, Sulfite (AutoCAT); Food and beverage: SO₂ in wine.
Application packages for use with TitraLab 1000 Series titrators from Hach contain all elements but reagents to make it quick and easy for anyone to set up and operate a test. Each application package is specifically designed for unique parameters to ensure accurate and reproducible measurements at glance. Packages include adapters, glass bottle, magnetic stirring bars, polypropylene beakers, bottle stopper with desiccant tube, electrode(s) (Digital IntelliCAL® or Analogue), high precision glass syringe (Hamilton) and USB key containing application notes and applications to automatically program titrators for selected parameters. Setting up your application has never been so simple.
Certifications: Safety IEC/EN 61010-1; EMC IEC/EN 61326-1
Accessories information: No applications kits or accessories included.