TitraLab® AT1000 Series Potentiometric Titrator, 1 Burette, 1 Pump, Hach
Supplier: Hach
AP0009.AT1112 AP0010.AT1112 AP0003.AT1112 AT1112.97
10662-160EA 2087.51 CAD
10662-160 10662-162 10662-168 10662-170 10662-240 10662-254 CA10662-160 CA10662-162 CA10662-168 CA10662-254
TitraLab® AT1000 Series Potentiometric Titrator, 1 Burette, 1 Pump, Hach
Titrators
Automatic titration without all the complications.
  • Reliable titration results
  • Eliminates operator interpretation and manual processes
  • Simple setup and titration
  • Application-specific functions
Simplify complex titration with the TitraLab® AT1000 from Hach.
Eliminates operator interpretation and manual processes with automatic titration to quickly deliver accurate and repeatable results. Pre-programmed titration methods detect end points and eliminate manual calculations to make results easier to achieve without advanced programming. Application-More Product Information